missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2C1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13658-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a nuclear hormone receptor characterized by a highly conserved DNA binding domain (DBD), a variable hinge region, and a carboxy-terminal ligand binding domain (LBD) that is typical for all members of the steroid/thyroid hormone receptor superfamily. This protein also belongs to a large family of ligand-inducible transcription factors that regulate gene expression by binding to specific DNA sequences within promoters of target genes. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these variants has not been determined.Specifications
NR2C1 | |
Polyclonal | |
Unconjugated | |
NR2C1 | |
NR2C1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22025, 252924, 7181 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13056, Q505F1, Q8VIJ4 | |
NR2C1 | |
NR2C1 Fusion Protein Ag4554 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |