missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15767-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
NQO2.Spécification
NQO2 | |
Polyclonal | |
Unconjugated | |
NQO2 | |
DHQV, DIA6, NQO2, NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, QR2, Quinone reductase 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18105, 4835 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16083, Q9JI75 | |
Nqo2 | |
NQO2 Fusion Protein Ag8403 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |