Learn More
NQO1 Mouse anti-Human, Unconjugated, Clone: 1E5G7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67240-1-IG-20UL
 View more versions of this product
Product Code. 16809765
  • 1710.33 NOK / 20µL
Description
The NQO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NQO1. This antibody recognizes human, rat, mouse, pig antigen. The NQO1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
NQO1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, ELISA
1E5G7
Unconjugated
20 μL
1728
IgG2a