Learn More
NQO1 Mouse anti-Human, Unconjugated, Clone: 1E5G7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67240-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16899755
  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
Description
The NQO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NQO1. This antibody recognizes human, rat, mouse, pig antigen. The NQO1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
NQO1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, ELISA
1E5G7
Unconjugated
150 μL
1728
IgG2a