missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPTX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10889-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the family of neuronal petraxins, synaptic proteins that are related to C-reactive protein. This protein is involved in excitatory synapse formation. It also plays a role in clustering of alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA)-type glutamate receptors at established synapses, resulting in non-apoptotic cell death of dopaminergic nerve cells. Up-regulation of this gene in Parkinson disease (PD) tissues suggests that the protein may be involved in the pathology of PD.Specifications
| NPTX2 | |
| Polyclonal | |
| Unconjugated | |
| NPTX2 | |
| NARP, Neuronal pentraxin 2, neuronal pentraxin II, NPTX2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288475, 4885, 53324 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O70340, P47972, P97738 | |
| Nptx2 | |
| NPTX2 Fusion Protein Ag1326 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction