Learn More
Cytiva nProtein A Sepharose™ 4 Fast Flow Affinity Media
Product Code. 11359931 Shop All Cytiva Products
Click to view available options
Quantity:
5 mL
25 mL
pH Range:
2 to 10
3 to 9
Unit Size:
25mL
5mL
Product Code. 11359931
Brand: Cytiva 17528001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used for purification of monoclonal and polyclonal antibodies. Cytiva Lifescience™ nProtein A Sepharose™ 4 Fast Flow Affinity Media contains native protein A coupled to Sepharose 4 Fast Flow.
  • Low leakage of protein A with nearly twice total IgG binding capacity of Protein A Sepharose CL-4B
  • Used in large-scale FDA-approved processes
  • Manufactured without using animal-derived components
  • Rigidity and high degree of substitution of Sepharose 4 Fast Flow matrix facilitates speedy process of large volumes dilute cell culture fluid
  • High chemical, thermal and mechanical stability
  • Withstands high concentrations of hydrogen bond disrupting agents
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 mL
Stability No significant change in chromatographic performance after 1 week storage using 8 M Urea, 6 M Gua-HCI, 2% Benzyl Alcohol or 20% Ethanol
For Use With (Application) Purification of Monoclonal and Polyclonal Antibodies
Storage Requirements 2°C to 8°C
Binding Capacity >35 mg Human IgG/mL
Product Type nProtein A Sepharose 4 Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 90 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 3 to 9
Storage Buffer 20% Ethanol
Ligand Type Native Protein A (S. aureus)