missing translation for 'onlineSavingsMsg'
Learn More

NPHS1 Monoclonal Antibody (OTI1G6), TrueMAB™, OriGene

Product Code. p-7225414
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17518274 30 μL 30µL
17598264 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17518274

Brand: Origene Technologies TA813411S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.

NPHS1 Monoclonal antibody specifically detects NPHS1 in Human samples. It is validated for Western Blot

Specifications

Antigen NPHS1
Applications Western Blot
Classification Monoclonal
Clone OTI1G6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Nphs1
Gene Accession No. O60500
Gene Alias CNF; Nephrin; nephrin 1; NephrinB; nephrosis 1 homolog, nephrin; nephrosis 1, congenital, Finnish type (nephrin); nephrosis 1, nephrin; NPHN; NPHS1; NPHS1 nephrin; NPHS1, nephrin; renal glomerulus-specific cell adhesion receptor
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1077-1241 of human NPHS1 (NP_004637) produced in E.coli.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4868
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.