missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NPHS1 (Human) Recombinant Protein
Human NPHS1 partial ORF ( NP_004637.1, 33 a.a. - 122 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004868-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: GFWALPENLTVVEGASVELRCGVSTPGSAVQWAKDGLLLGPDPRIPGFPRYRLEGDPARGEFHLHIEACDLSDDAEYECQVGRSEMGPEL
Specifications
NP_004637.1 | |
nephrosis 1, congenital, Finnish type (nephrin) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NPHS1 | |
GST |
4868 | |
Wheat germ expression system | |
10 μg | |
CNF, NPHN | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |