Learn More
Thermo Scientific Chemicals NP-40 lysis buffer
Product Code. p-7041936
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. 15403489
Brand: Thermo Scientific Chemicals J60766.AP
Request Bulk or Custom Format
Edge
NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.
Solubility
It is soluble in water.
Notes
This buffer contains: 50mM Tris-HCl (pH 7.4), 150mM NaCl, 1%NP-40 and 5mM EDTA. To use, incubate cells in 5 volumes of buffer on ice for 10-30 minutes.
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
Content And Storage Keep cold
Format Liquid
Quantity 500 mL