missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOX5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25350-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is predomitly expressed in the testis and lymphocyte-rich areas of spleen and lymph nodes. It encodes a calcium-dependen NADPH oxidase that generates superoxide, and functions as a calcium-dependent proton channel that may regulate redox-dependent processes in lymphocytes and spermatozoa. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
| NOX5 | |
| Polyclonal | |
| Unconjugated | |
| NOX5 | |
| NADPH oxidase 5, NOX5, NOX5A, NOX5B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79400 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96PH1 | |
| NOX5 | |
| NOX5 Fusion Protein Ag18673 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction