missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human RPS2 His Protein

Product Code. 18710564 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18710564 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18710564 Supplier Novus Biologicals™ Supplier No. NBP223433

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-293 of Human RPS2 The Recombinant Human RPS2 Protein is derived from E. coli. The Recombinant Human RPS2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_002943
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.4M UREA
Gene ID (Entrez) 6187
Molecular Weight (g/mol) 33.7kDa
Name RPS2 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen RPS2, 1-293aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMADDAGA AGGPGGPGGP GMGNRGGFRG GFGSGIRGRG RGRGRGRGRG RGARGGKAED KEWMPVTKLGRLVKDMKIKS LEEIYLFSLP IKESEIIDFF LGASLKDEVL KIMPVQKQTR AGQRTRFKAF VAIGDYNGHV GLGVKCSKEV ATAIRGAIIL AKLSIVPVRR GYWGNKIGKP HTVPCKVTGR CGSVLVRLIP APRGTGIVSA PVPKKLLMMA GIDDCYTSAR GCTATLGNFA KATFDAISKTYSYLTPDLWK ETVFTKSPYQ EFTDHLVKTH TRVSVQRTQA PAVATT
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.