missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PRL-3/PTP4A3 His Protein

Product Code. 18291270 Shop All Bio Techne Products
Change view
Click to view available options
:
0.1mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18291270 0.1mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18291270 Supplier Novus Biologicals™ Supplier No. NBP130246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-173 of Human PRL-3/PTP4A3 The Recombinant Human PRL-3/PTP4A3 Protein is derived from E. coli. The Recombinant Human PRL-3/PTP4A3 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_116000
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 2mM EDTA, 1mM DTT, 10% glycerol
Gene ID (Entrez) 11156
Molecular Weight (g/mol) 21.6kDa
Name PRL-3/PTP4A3 Protein
Purification Method Protein
Immunogen MGSSHHHHHH SSGLVPRGSH MARMNRPAPV EVSYKHMRFL ITHNPTNATL STFIEDLKKY GATTVVRVCE VTYDKTPLEK DGITVVDWPF DDGAPPPGKV VEDWLSLVKA KFCEAPGSCV AVHCVAGLGR APVLVALALI ESGMKYEDAI QFIRQKRRGA INSKQLTYLE KYRPKQRLRF KDPHTHKTRC CVM
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.