missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-270 of uman MED4 The Recombinant Human MED4 Protein is derived from E. coli. The Recombinant Human MED4 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | AAH05189 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 20% glycerol, 1mM DTT, 100mM NaCl |
| Gene ID (Entrez) | 29079 |
| Molecular Weight (g/mol) | 30.7kDa |
| Name | MED4 Protein |
| Purification Method | Protein |
| Immunogen | MAASSSGEKE KERLGGGLGV AGGNSTRERL LSALEDLEVL SRELIEMLAI SRNQKLLQAG EENQVLELLI HRDGEFQELM KLALNQGKIH HEMQVLEKEV EKRDGDIQQL QKQLKEAEQI LATAVYQAKE KLKSIEKARK GAISSEEIIK YAHRISASNA VCAPLTWVPG DPRRPYPTDL EMRSGLLGQM NNPSTNGVNG HLPGDALAAG RLPDVLAPQY PWQSNDMSMN MLPPNHSSDF LLEPPGHNKE DEDDVEIMST DSSSSSSESD LEHHHHHH |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?