Learn More
Novus Biologicals™ A-431 Whole Cell Lysate
Brand: Novus Biologicals™ NB820-59461
This item is not returnable.
View return policy
Description
The A-431 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.Specifications
| Immunoprecipitation, Western Blot | |
| 1 mg | |
| Protein | |
| Whole | |
| -80°C. Avoid freeze-thaw cycles. |
| Skin | |
| HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
| Total Protein - Human Tumor Cell Line: A431 | |
| Human | |
| Native |