Learn More
Novus Biologicals™ A-431 Whole Cell Lysate
Brand:  Novus Biologicals™ NB820-59461
Encompass
Product Code. 15758095
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The A-431 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Immunoprecipitation, Western Blot
1 mg
Protein
Whole
-80°C. Avoid freeze-thaw cycles.
Skin
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Total Protein - Human Tumor Cell Line: A431
Human
Native
Certificates
Promotions
Promotions
Product Title
Select an issue
For Research Use Only