Learn More
Invitrogen™ Novex™ Value™ Tris-Glycine Mini Protein Gel, 4 to 12%, 1.0 mm, 15-well
Product Code. 15653686
Change view
Click to view available options
Quantity:
20 gels (1 box)
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15653686 20 gels (1 box) Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15653686 Supplier Invitrogen™ Supplier No. XV04125PK20
Includes
20 precast gels
Separation Range: 20 to 250 kd; Separation Type: Denaturing, Native; For Use With: XCell SureLock Mini
Features of the Novex Value gels include:
  • Great balance of value and performance
  • Fast run conditions - separate your proteins using constant voltage in less than 60 minutes
  • Improved shelf life - store gels for at least 12 months at 4°C
  • Usable for native and denaturing protein assays
Warranty
Up to 12 months
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in refrigerator (2–8°C).
Shelf life up to 12 months depending on gel type.
Description 4-12% Value Tris-Glycine Gels, 1.0 mm, 15-well, Box of 20
Gel Percentage 4 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine
Recommended Applications Denaturing, Native
Sample Loading Volume up to 15 μL
Separation of Protein
Separation Range 20 to 250 kDa
Separation Type Denaturing, Native
Wells 15-well
For Use With (Application) Protein Gel Electrophoresis
Material Tris-Glycine Gel
Mode of Separation Molecular Weight
Quantity 20 gels (1 box)
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Storage Requirements Store at 4°C. Shelf life is 12 months.
Thickness 1.0 mm
Well Design 1D Well
Length (Metric) 8 cm
Length (Metric) Gel 8 cm
Width (Metric) 8 cm
Width (Metric) Gel 8 cm
Product Line Novex Value
Product Type Gradient Page
For Use With (Equipment) XCell SureLock Mini-Cell