Learn More
Invitrogen™ Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675
Description
Includes
Novex Tricine SDS Running Buffer (10X, 500mL - LC1675) and Novex Tricine SDS Sample Buffer (2X, 20mL - LC1676)
In the tricine system developed by Schaegger and von Jagow (1), tricine replaces glycine in the running buffer. This results in more efficient stacking and destacking of low molecular weight proteins and higher resolution of smaller peptides.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Product Type | Buffer Kit |
| Content And Storage | The Novex™ Tricine SDS Buffer Kit includes Novex™ Tricine SDS Running Buffer (10X, 500 ml - LC1675) and Novex™ Tricine SDS Sample Buffer (2X, 20 ml - LC1676). |
| Gel Type | SDS PAGE Gel |
| Gel Compatibility | Novex Tricine Gel |
| Product Line | Novex |
| Quantity | 1 kit |