Learn More
Invitrogen™ StainEase™ Staining Trays
Description
Includes
StainEase™ Gel Staining Tray comes with two clear polycarbonate containers, two white polycarbonate strainers, two clear polycarbonate lids with handles and waterproof lid labels.
The StainEase Gel Staining Tray is a convenient way to evenly stain mini-gels and prevent gels from breaking. The staining tray assembly includes a perforated strainer insert that holds the gel and drains away the stain, a lid, and an outer container. Easily fits up to four mini-gels.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Contents: StainEase™ Gel Staining Tray comes with two clear polycarbonate containers, two white polycarbonate strainers, two clear polycarbonate lids with handles and waterproof lid labels. |
| Quantity | 2 pack |
| Shipping Condition | Room Temperature |
| Product Line | StainEase |
| Product Type | Staining Tray |