Learn More
Invitrogen™ Novex™ pH 3-10 IEF Buffer Kit,Includes LC5300, LC5310, LC5311
Product Code. 10297142
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Product Code. Quantity unitSize
10297142 1 kit 1 set
1 options
Product Code. 10297142
Brand: Invitrogen™ LC5317
Includes
Each kit includes Novex IEF Anode Buffer, 50X (LC5300), Novex IEF Cathode Buffer pH 3-10, 10X (LC5310), Novex IEF Sample Buffer pH 3-10, 2X (LC5311)
Optimized for using with Novex™ pH 3-10 IEF gels
  • Concentration: 20X
Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Buffer Anode & Cathode Buffers
Concentration 20 X
Gel Type IEF
pH Range 3 to 10
Product Type IEF Buffer Kit
Gel Compatibility Novex™ IEF Gels
Product Line Novex
Quantity 1 kit