missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ 4 to 20%, Tris-Glycine, 1.0 mm, Mini Protein Gel, 15-well

Product Code. 12015685
Click to view available options
Wells:
15-well
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12015685

Brand: Invitrogen™ EC60255BOX

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

One box of 10 gels

Separation Range: 20 to 200 kd; Separation Type: Denaturing, Native; For Use With: Mini Gel Tank, XCell SureLock Mini

This product is being replaced by Catalog number XP04205BOX and will no longer be available after May 31, 2016.

  • Provide reproducible separation of a wide range of proteins into well-resolved bands
  • Individually packaged gels for optimal experiment design
  • Compatible with most protein standards; usable for native and denaturing protein assays
  • For best results, most Novex Tris-Glycine gels should be stored at 4°C and used within 8 weeks of purchase
  • Run proteins in native or denatured form
  • Do not contain SDS and can be used to run proteins in native or in denatured form

Sample Loading Volume: 15μL; 1D Well

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) XCell SureLock Mini-Cell
Gel Percentage 4 to 20%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine
Product Type Gradient Page
Separation of Protein
Separation Range 20 to 200 kDa
Separation Type Denaturing, Native
Wells 15-well
Quantity Each
Well Design 1D Well
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.