missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Novex™ 10%, Tris-Glycine, 1.0 mm, Midi Protein Gels, 12+2-well, w/adapters

Product Code. 16336585
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.

Product Code. 16336585

Brand: Novex™ WXP01012BOXA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.

Includes

Adapters for BioRad Criterion Cell

Separation Range: 30 to 250 kd, Separation Type: Denaturing, Native, For Use With: XCell4 SureLock Midi, BioRad Criterion

Novex™ Tris-Glycine Plus Midi Protein gels are improved polyacrylamide gels based ontraditional Laemmli protein electrophoresis, which allows the use of Laemmli sample and running buffers. Novex Tris-Glycine Plus gels offer reproducible separation of a wide range of proteins into well-resolved bands. The Novex Tris-Glycine Plus Midi Protein Gels are a 1.0 mm thick, wider format (8 cm x 13 cm) gel for higher throughput electrophoresis of protein samples. The included adapter is designed for use with the BioRad Criterion™ Cell.

  •  Improved shelf life—store gels for up to 12 months at 4°C
  • Diversity—use for native and denaturing protein assays
  • Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Recommended Storage

Store in refrigerator (2-8°C). Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) XCell4 SureLock Midi-Cell, Bio-Rad Criterion Cell
Gel Percentage 10%
Gel Size Midi
Gel Thickness 1.0 mm
Gel Type Tris-Glycine
Separation of Protein
Separation Range 30 to 250 kDa
Separation Type Denaturing, Native
Wells 12 + 2-well, w/adapters
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.