Learn More
NOTUM Mouse anti-Human, Unconjugated, Clone: 2F6D5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66956-1-IG-20UL
 View more versions of this product
Product Code. 16809285
  • 1710.33 NOK / 20µL
Description
The NOTUM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NOTUM. This antibody recognizes human antigen. The NOTUM antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
NOTUM
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
2F6D5
Unconjugated
20 μL
147111
IgG1