missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notoginsenoside Fe, MedChemExpress
Description
Notoginsenoside Fe is a natural compound isolated from Panax japlcus var.
Specifications
Specifications
| Chemical Name or Material | Notoginsenoside Fe |
| CAS | 88105-29-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C47H80O17 |
| Quantity | 5 mg |
| Synonym | Notoginseng triterpenes Ginsenoside Mb |
| Solubility Information | H2O : 50 mg/mL (54.52 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble) |
| SMILES | C[C@]12[C@@](C)([C@]3([H])C[C@@H](O)[C@]1([H])[C@@]([H])([C@](C)(CC/C=C(C)/C)O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](CO)[C@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)CC2)CC[C@@]([C@]3(C)CC6)([H])C(C)(C)[C@H]6O[C@@]7([H])[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O7 |
| Molecular Weight (g/mol) | 917.13 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?