missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NOTCH2NL Recombinant Protein

Recombinant protein for NOTCH2NL (human) gene

Brand:  Abnova™ H00388677-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190494

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Human NOTCH2NL full-length ORF ( AAH19835, 1 a.a. - 236 a.a.) recombinant protein with GST-tag at N-terminal
  • Theoretical molecular weight: 51.7kDa
  • Preparation: in vitrowheat germ expression system
  • Purification: glutathione sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer

Sequence: MCVTYHNGTGYCKCPEGFLGEYCQHRDPCEKNRCQNGGTCVAQAMLGKATCRCASGFTGEDCQYSTSHPCFVSRPCLNGGTCHMLSRDTYECTCQVGFTGKECQWTDACLSHPCANGSTCTTVANQFSCKCLTGFTGQKCETDVNECDIPGHCQHGGICLNLPGSYQCQCLQGFTGQY CDSLYVPCAPSPCVNGGTCRQTGDFTFECNCLPETVRRGTELWERDREVWNGKEHDEN

Best use within three months from the date of receipt.

ELISA, western blotting (recombinant protein), antibody production, protein array

Specifications

Specifications

AAH19835
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
51.7
8
Glutathione Sepharose 4 Fast Flow
25 ug
MCVTYHNGTGYCKCPEGFLGEYCQHRDPCEKNRCQNGGTCVAQAMLGKATCRCASGFTGEDCQYSTSHPCFVSRPCLNGGTCHMLSRDTYECTCQVGFTGKECQWTDACLSHPCANGSTCTTVANQFSCKCLTGFTGQKCETDVNECDIPGHCQHGGICLNLPGSYQCQCLQGFTGQYCDSLYVPCAPSPCVNGGTCRQTGDFTFECNCLPETVRRGTELWERDREVWNGKEHDEN
N2N
NOTCH2NL
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
388677
NOTCH2NL (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NOTCH2NL
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers