missing translation for 'onlineSavingsMsg'
Learn More

Notch1 Mouse anti-Human, Oligo, Clone: MHN1-519, BD™ AbSeq ™

Product Code. 17273335
Change view
Click to view available options
Quantity:
25 Tests
Unit Size:
25 tests
Product Code. Quantity unitSize
17273335 25 Tests 25 tests
1 options
This item is not returnable. View return policy

Product Code. 17273335

Brand: BD Biosciences 940292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Notch1 Monoclonal antibody specifically detects Notch1 in Human samples. It is validated for Single Cell 3' Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen Notch1
Applications Single Cell 3' Sequencing
Classification Monoclonal
Clone Clone MHN1-519
Conjugate Oligo
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias NOTCH1; Notch 1; NOTC1; hN1; TAN1; Neurogenic locus notch homolog protein 1
Host Species Mouse
Immunogen Human Notch1 Recombinant Protein
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.