Learn More
Honeywell™ North™ NorthFlex Cold Grip™ PVC-Coated Polyamide Insulated Gloves
Click to view available options
Size:
Large
X-Large
Unit Size:
12 pairs
Description
- Heavyweight insulated gloves without sacrificing comfort and dexterity
- Polyamide yarn reduces linting for the sensitive applications
- Black PVC coating covers the palm and fingers for enhanced grip capabilities
- Hydropellent Technology repels liquids to provide a dry, firm grip
- Ideal for handling damp, oily, or greasy parts in cold conditions
- Recommended for temperatures down to -26°C (-15°F)
- Launderable
Specifications
Specifications
| Coating Material | PVC |
| Color | Black |
| Exterior Finish | Fully Textured |
| Hand | Specific |
| Material | Polyamide |
| Size | Large |
| For Use With (Application) | Construction, Logistics, and Maintenance |
| Glove Cuff Color | Blue |
| Glove Cuff Style | Knit |
| Liner Material | Synthetic Insulation |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering