missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norrin/NDP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
2650.00 NOK - 4900.00 NOK
Specifications
| Antigen | Norrin/NDP |
|---|---|
| Dilution | Western Blot Reactivity reported in (PMID: 29860944)., Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18400701
|
Novus Biologicals
NBP1-84769-25ul |
25ul |
2650.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18705263
|
Novus Biologicals
NBP1-84769 |
4900.00 NOK
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Norrin/NDP Polyclonal specifically detects Norrin/NDP in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Norrin/NDP | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EVR2, exudative vitreoretinopathy 2 (X-linked), FEVR, ND, Norrie disease (pseudoglioma), Norrie disease protein, norrin, X-linked exudative vitreoretinopathy 2 protein | |
| NDP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot Reactivity reported in (PMID: 29860944)., Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, Neuroscience, Vision | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 4693 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TDSSFIMDSDPRRCMRHHYVDSISHPLYKCSSKMVLLARCEGHCSQASRSEPLVSFSTVLKQPFRSSCHCCRPQTSKLKALRLRCSGGMRLTATYRYILSCHCEECNS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto