Learn More
Norrin/NDP Antibody, Novus Biologicals™
Product Code. 18705263 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.10mL
25µL
Quantity:
25ul
Product Code. unitSize Quantity
18705263 0.10mL -
18400701 25µL 25ul
2 options
Product Code. 18705263
Brand: Novus Biologicals NBP184769
Rabbit Polyclonal Antibody has been used in 1 publication
Norrin/NDP Polyclonal specifically detects Norrin/NDP in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Dilution Western Blot Reactivity reported in (PMID: 29860944)., Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Gene Alias EVR2, exudative vitreoretinopathy 2 (X-linked), FEVR, ND, Norrie disease (pseudoglioma), Norrie disease protein, norrin, X-linked exudative vitreoretinopathy 2 protein
Gene Symbols NDP
Purification Method Affinity Purified
For Research Use Only