Learn More
Masterflex™ L/S™ Norprene™ High-Pressure Pump Tubing
Click to view available options
Tubing Size:
15
16
Unit Size:
Each
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
Spécification
Spécification
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 4.8 mm |
| Material | Norprene |
| Overall Length (Metric) | 7.5 m |
| For Use With | L/S high-performance pump head models 77250-62 and 77250-82 only |
| For Use With (Application) | high pressure applications |
| Tubing Size | 15 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu