Learn More
Invitrogen™ RNase-free Microfuge Tubes
Product Code. 10499034
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Surface Type:
Graduated
Graduated, low-binding
Quantity:
1000 Tubes/Pk.
250 Tubes/Pk.
500 Tubes/Pk.
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
Product Code. Capacity (Metric) Surface Type Quantity unitSize
10499034 0.5 mL Graduated, low-binding 1000 Tubes/Pk. Pack of 1000
10259544 0.5 mL Graduated, low-binding 500 Tubes/Pk. Pack of 500
10676825 1.5 mL Graduated, low-binding 250 Tubes/Pk. Pack of 250
10409044 1.5 mL Graduated, low-binding 500 Tubes/Pk. Pack of 500
10318574 2 mL Graduated, low-binding 250 Tubes/Pk. Pack of 250
10371235 2 mL Graduated 500 Tubes/Pk. Pack of 500
6 options
Product Code. 10499034
Brand: Invitrogen™ AM12300
Only null left
Add to basket
Edge
Add to basket
These microcentrifuge tubes are guaranteed RNase- and DNase-free.
Invitrogen microcentrifuge tubes are guaranteed RNase- and DNase-free.
Features include:
  • Marked with graduations for convenience
  • Flat lids for easy labeling
  • Positive seal design that prevents caps from opening in boiling water baths
  • Autoclave-safe
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Capacity (Metric) 0.5 mL
Material Polypropylene
Quantity 1000 Tubes/Pk.
Content And Storage Store at room temperature.
Product Line Invitrogen
Shipping Condition Room Temperature
Dnase Free RNase-Free, DNase-Free
Autoclavable Autoclavable
Closure Type Snap Cap
Closure Included Yes
Graduated Yes
Product Type Microfuge Tube (RNase-free)
Surface Type Graduated, low-binding