Learn More
Nonanoic Acid, Sigma-Aldrich™
Product Code. 15889430
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15889430 100 mL 100mL
15856440 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15889430 Supplier Sigma Aldrich Supplier No. 8071670100
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 112-05-0
Specifications
Melting Point 13°C
Density 0.91 g/cm3 at 20°C
Molecular Formula CH3(CH2)7COOH
MDL Number MFCD00004433
Quantity 100 mL
Synonym Nonanoic acid, Pelargonic acid, Nonanoic acid
Molecular Weight (g/mol) 158.24
Grade Research