Learn More
Nonanoic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Nonanoic-2,2-D2 Acid |
| CAS | 62689-94-5 |
| Molecular Formula | C9 D2 H16 O2 |
| Quantity | 1 g |
| Synonym | Nonanoic-2,2-d2 acid (9 CI, ACI), α-Dideuterononanoic acid, Nonanoic-2,2-D2 acid, 1-Octanecarboxylic-1,1-d2 acid, n-Nonanoic acid-d2, Pelargonic acid-α-d2, Nonanoic acid-d2 |
| InChI Formula | InChI=1 S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)/i8D2 |
| SMILES | [2 H]C([2 H])(CCCCCCC)C(=O)O |
| IUPAC Name | 2,2-dideuteriononanoic acid |
| Molecular Weight (g/mol) | 160.25 |
| Formula Weight | 160.143 g/mol |
| Show More |