Learn More
Thermo Scientific™ Non-pressurized Plastic Water Deionizer Replacement Cartridges
These non-pressurized replacement cartridges provide deionized water for many applications and are for use with Thermo Scientific™ Non-pressurized Plastic Water Deionizer.
Brand: Thermo Scientific™ 50155437
Additional Details : Weight : 1.68300kg
Description
Thermo Scientific™ Non-pressurized Plastic Water Deionizer replacement cartridges provide deionized water for many applications. For use with Non-Pressurized Plastic Water Deionizers, small compact systems that provide demineralized water. Ideal for small laboratories but also for supplying autoclaves and washers in doctors' and dentists' practices.
Specifications
Replacement Cartridge for DI 425 | |
425 L | |
0.1 to 20 μS/cm |
50 L/hr. | |
DI 425 pressureless (Part Number 01.0425) | |
Non-pressureized Deionizer |