missing translation for 'onlineSavingsMsg'
Learn More

NOL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14802-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893624

  • 3468.15 NOK / 150µL
Estimated Shipment: 13-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NOL4
Polyclonal
Unconjugated
NOL4
HRIHFB2255, NOL4, NOLP, Nucleolar localized protein, nucleolar protein 4
Rabbit
Antigen Affinity Chromatography
RUO
307553, 319211, 8715
-20°C
Liquid
Western Blot
0.44 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O94818, P60954
Nol4
NOL4 Fusion Protein Ag6518
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers