missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14456-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
In yeast, over 200 protein and RNA cofactors are required for ribosome assembly, and these are generally conserved in eukaryotes. These factors orchestrate modification and cleavage of the initial 35S precursor rRNA transcript into the mature 18S, 5.8S, and 25S rRNAs, folding of the rRNA, and binding of ribosomal proteins and 5S RNA. Nob1 is involved in pre-rRNA processing. In a late cytoplasmic processing step, Nob1 cleaves a 20S rRNA intermediate at cleavage site D to produce the mature 18S rRNA.Specifications
NOB1 | |
Polyclonal | |
Unconjugated | |
NOB1 | |
ART 4, ART4, MST158, MSTP158, NOB1, NOB1P, Protein ART 4, PSMD8BP1, RNA binding protein NOB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28987, 291996, 67619 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6VEU1, Q8BW10, Q9ULX3 | |
NOB1 | |
NOB1 Fusion Protein Ag6051 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |