Learn More
Abnova™ NME1 (Human) Recombinant Protein
Description
- Sequence: MANCERTFIAIKPDGVQRGLVGEIIKRFEQKGFRLVGLKFMQASEDLLKEHYVDLKDRPFFAGLVKYMHSGPVVAMVWEGLNVVKTGRVMLGETNPADSKPGTIRGDFCIQVGRNIIHGSDSVESAEKEIGLWFHPEELVDYTSCAQNWIYE
Specifications
Specifications
| Accession Number | AAH00293 |
| Gene ID (Entrez) | 4830 |
| Name | non-metastatic cells 1, protein (NM23A) expressed in |
| Preparation Method | Wheat germ expression system |
| Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | AWD, GAAD, NB, NBS, NDPK-A, NDPKA, NM23, NM23-H1 |
| Gene Symbol | NME1 |
| Species | Wheat Germ (in vitro) |
| Show More |