Learn More
NLN Monoclonal Antibody (OTI3H4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA525881
Additional Details : Weight : 0.01000kg
Description
This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.Specifications
NLN | |
Monoclonal | |
0.92 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42676, Q91YP2, Q9BYT8 | |
Nln | |
Full length human recombinant protein of NLN produced in HEK293T cell | |
100μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3H4 | |
Unconjugated | |
Nln | |
4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN | |
Mouse | |
Affinity Chromatography | |
RUO | |
117041, 478081, 57486, 75805 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |