missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKIRAS1 Antibody (184C278) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-56555JF635
This item is not returnable.
View return policy
Description
NKIRAS1 Monoclonal antibody specifically detects NKIRAS1 in Bovine,Canine,Equine,Human,Mouse,Porcine,Primate,Rat,Xenopus samples. It is validated for Western Blot
Specifications
| NKIRAS1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| I-kappa-B-interacting Ras-like protein 1, kappa B-ras 1, Kappa B-Ras protein 1, KappaB-Ras1, KBRAS1kappaB-Ras1, NF-kappa-B inhibitor-interacting Ras-like protein 1, NFKB inhibitor interacting Ras-like 1, NFKB inhibitor interacting Ras-like protein 1 | |
| This monoclonal antibody was raised against a synthetic peptide containing amino acid residues 96-113 (RVELLKKEIDKFKDKKEV) of human kB-Ras1. | |
| 0.1 mL | |
| Signal Transduction | |
| 28512 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot | |
| 184C278 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Equine, Human, Mouse, Pig, Primate, Rat, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction