missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKG2A/CD159a/KLRC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17084
This item is not returnable.
View return policy
Description
NKG2A/CD159a/KLRC1 Polyclonal specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for Western Blot.
Specifications
| NKG2A/CD159a/KLRC1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
| Rabbit | |
| 26 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 3821 | |
| Human | |
| IgG |
| Western Blot | |
| 0.2mg/mL | |
| Western Blot 1:500-1:3000 | |
| P26715 | |
| KLRC1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human NKG2A. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Rhesus Monkey (100%), Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction