Learn More
NK-252, MedChemExpress
Product Code. 16493817
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16493817
Brand: Medchem Express HY19734
MedChemExpress NK-252 is a potential Nrf2 activator, which exhibits a great Nrf2-activating potential.
NK-252 is a potential Nrf2 activator, which exhibits a great Nrf2-activating potential.
Specifications
Chemical Name or Material NK-252
Physical Form Solid
CAS 1414963-82-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C13H11N5O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 29 mg/mL (101.66 mM)
SMILES O=C(NCC1=NC=CC=C1)NC2=NN=C(C3=CC=CO3)O2
Molecular Weight (g/mol) 285.26
Formula Weight 285.26
Percent Purity 99.48%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.