Learn More
Nizatidine Sulfoxide, TRC
Product Code. 30471287
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471287 25 mg 25mg
30476730 100 mg -
30473773 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30471287 Supplier Toronto Research Chemicals Supplier No. TRCN59851025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102273-13-2
Molecular Formula C12 H21 N5 O3 S2
Molecular Weight (g/mol) 347.46
Synonym (EZ)-N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine, Nizatidine Imp. C (EP)
IUPAC Name 1-N'-[2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfinyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine
SMILES CNC(=C[N+](=O)[O-])NCCS(=O)Cc1csc(CN(C)C)n1
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C12H21N5O3S2/c1-13-11(6-17(18)19)14-4-5-22(20)9-10-8-21-12(15-10)7-16(2)3/h6,8,13-14H,4-5,7,9H2,1-3H3
Formula Weight 347.1086
Chemical Name or Material (EZ)-N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine