Description
- Nitrile coating covers the fingers and the knuckles for an effective barrier against oil and grease
- 100% cotton fleeced interlock liner for increased comfort
- Stronger and more flexible than PVC
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 5 |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, REACH Compliant |
| Coating Material | Nitrile |
| Cut Resistance Rating | ANSI A2 |
| Hand | Specific |
| Size | 8 |
| EN388 Standard | 4221B |
| For Use With (Application) | Automotive Assembly and Metal Fabrication |
| Glove Cuff Color | Natural |
| Glove Cuff Style | Knit |
| Show More |