Learn More
Ansell NITROTOUGH™ N250Y Medium Duty Oil-Repellant Gloves
Catalog Number 16865310
Click to view available options
Size:
7
8
9
10
11
Nitrile coating all over the gloves provides a high level of oil repellance, while the elasticated knit wrist ensures a secure fit.
  • Nitrile coating provides oil repellence on the palm and back of the hand
  • Elasticated knitwrist ensures a secure fit
  • Comfortable and flexible
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Coating Material Nitrile
Color Yellow
Size 7
EN388 Standard 4111A
For Use With (Application) Assembly, General Maintenance, Metal Fabrication, and Quality Control
Glove Cuff Color Natural
Glove Cuff Style Knit
Liner Material 100% Cotton
Series N250Y