Learn More
Ansell NITROTOUGH™ N250Y Medium Duty Oil-Repellant Gloves
Click to view available options
Size:
7
8
9
10
11
Description
- Nitrile coating provides oil repellence on the palm and back of the hand
- Elasticated knitwrist ensures a secure fit
- Comfortable and flexible
Specifications
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
Coating Material | Nitrile |
Color | Yellow |
Size | 9 |
EN388 Standard | 4111A |
For Use With (Application) | Assembly, General Maintenance, Metal Fabrication, and Quality Control |
Glove Cuff Color | Natural |
Glove Cuff Style | Knit |
Liner Material | 100% Cotton |
Series | N250Y |