missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, 100.0%, 99.0% purity, For GC analysis, MP Biomedicals™

Product Code. p-4405155 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Product Code. Quantity unitSize
11459121 500 mL 500mL
11469121 1 L 1 L
2 options
This item is not returnable. View return policy

Product Code. 11459121

Brand: MP Biomedicals 0215585190

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym benzene, nitro, nitro-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j
PubChem CID 11473
ChEBI CHEBI:27986
IUPAC Name nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1

Specifications

Color Yellow
Density 1.196g/mL at 25°C
Assay Percent Range 99%
Quantity 500 mL
Formula Weight 123.12
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material Nitrobenzene, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.