Learn More
Meditrade™ Nitril™ NextGen™ Nitrile Examination Gloves
Product Code. 16671582
Click to view available options
Size:
Small
Medium
Large
X-Large
Color:
Blue
Unit Size:
Pack of 100
Product Code. 16671582
Brand: Meditrade™ 1283M
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Non-sterile Meditrade™ disposable nitrile examination gloves provide excellent protection for environments with a high risk of contamination. Powder-free and latex-free they are ideal for use in clinical, laboratory, and manufacturing environments.
  • Disposable examination glove
  • Nitrile
  • Non-sterile
  • Powder-free
  • Latex-free
  • Textured
  • Beaded cuff
  • Ambidextrous
  • AQL 1.5
Specifications
Certifications/Compliance AQL 1.5; EN ISO 13485:2016; 93/42/EWG / EEC: Class 1; PPE Category III; Food Compliance acc. to EC Directive 1935/2004 (blue only)
Description Single use disposable blue nitrile gloves
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
Size Medium
Thickness (Metric) 0.060 mm (Cuff)
Model 1283M