Learn More
Tocris Bioscience™ NITD008
Antiviral; selective flavivirus inhibitor
Brand: Tocris Bioscience™ 6045/1
This item is not returnable.
View return policy
Description
NITD 008 is a selective flavivirus inhibitor. Adenosine analog. Inhibits dengue virus (EC50 = 0.64 μM), hepatitis C virus and cellular entry of Zika virus. Reduces increases of cytokines and NS1 and prevents death in dengue-infected mice. Active in vivo. Orally bioavailable.Specifications
| 7-(2-C-Ethynyl-β-D-ribofuranosyl)-7 H-pyrrolo[2,3-d]pyrimidin-4-amine | |
| 1044589-82-3 | |
| >98% | |
| 1 mg |