Learn More
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets
Product Code. 15392265
Change view
Click to view available options
pH:
10.012 (at 25°C)
4.006 (at 25°C)
6.865 (at 25°C)
9.180 (at 25°C)
Unit Size:
60mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Unit Size
15392265 4.006 (at 25°C) Each
15312275 6.865 (at 25°C) Each
15322275 9.180 (at 25°C) Each
17803775 10.012 (at 25°C) 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15392265 Supplier Mettler Toledo™ Supplier No. 30111136
  • ONLINE EXCLUSIVE PRICE
    1700.00 NOK
    1300.28 NOK / Pack of 1
    Save 399.72 NOK
    23% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Offers the highest possible pH precision. Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets are specified to three decimal places and come with a printed certificate that guarantees traceability. Individual packets guarantee fresh solution for every calibration, as well as maximum ease of use.
pH measurements are only as accurate as the buffer solutions used for calibration purposes. Maximum precision is guaranteed with fully traceable NIST/DIN buffers. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
  • NIST/DIN pH buffers are specified and guaranteed to three decimal places, offering top precision
  • Individual sachets guarantee fresh solution for every calibration, as well as maximum ease of use
  • A test certificate and detailed label with a temperature table comes with every box
  • All SDS and labels contain information according to GHS (Globally Harmonized System)
TRUSTED_SUSTAINABILITY
Specifications
Description NIST/DIN Buffer pH 4.006 sachets 30x20mL
For Use With (Application) pH measurement, electrode calibration
For Use With (Equipment) pH Electrode
pH 4.006 (at 25°C)
Quantity 30 x 20 mL
Packaging Sachet
Traceability to NIST Yes