Learn More
Nisoldipine, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Nisoldipine
Specifications
Specifications
| Chemical Name or Material | Nisoldipine |
| CAS | 63675-72-9 |
| Analyte or Component Names | Nisoldipine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 N2 O6 |
| Certifications/Compliance | ISO 17025 |
| InChI Formula | InChI=1S/C20H24N2O6/c1-11(2)10-28-20(24)17-13(4)21-12(3)16(19(23)27-5)18(17)14-8-6-7-9-15(14)22(25)26/h6-9,11,18,21H,10H2,1-5H3 |
| SMILES | COC(=O)C1=C(C)NC(=C(C1c2ccccc2[N+](=O)[O-])C(=O)OCC(C)C)C |
| IUPAC Name | 3-O-methyl 5-O-(2-methylpropyl) 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| Molecular Weight (g/mol) | 388.41 |
| Show More |