Learn More
Ninhydrin, EMD-Millipore
Product Code. 15824910
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15824910 100 mL Glass 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15824910 Supplier Supelco Supplier No. 1067050100
 Restricted Item
Estimated Shipment: 18-11-2026
Ready-to-use spray solutions for thin-layer chromatography
TRUSTED_SUSTAINABILITY
Specifications
CAS 485-47-2
Explosion Limit 2 to 13.4 %(V) (2-Propanol)
Ignition Point 425°C (2-Propanol)
Packaging Glass
Molecular Formula C9H6O4
Quantity 100 mL
Synonym ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
IUPAC Name 2,2-dihydroxyindene-1,3-dione
Molecular Weight (g/mol) 178.143
PubChem CID 10236
ChEBI CHEBI:86374
Vapor Pressure 42.4 hPa (20°C)
Chemical Name or Material Ninhydrin