Learn More
Ningetinib Tosylate, MedChemExpress
Product Code. 16490457
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490457 50 mg 50mg
16456807 5 mg 5mg
16435076 100 mg 100mg
16414108 10 mg 10mg
16402956 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16490457 Supplier Medchem Express Supplier No. HY107145
MedChemExpress Ningetinib Tosylate is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and c-Met, VEGFR2 and Axl, respectively.
Ningetinib Tosylate is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and c-Met, VEGFR2 and Axl, respectively.
Specifications
Chemical Name or Material Ningetinib Tosylate
CAS 1394820-77-9
Color White
Purity Grade Notes Research
Molecular Formula C38H37FN4O8S
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (45.73 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC=C(S(=O)(O)=O)C=C1.CC2=C(C(NC3=CC(F)=C(OC4=CC=NC5=C4C=CC(OCC(C)(O)C)=C5)C=C3)=O)C(N(C6=CC=CC=C6)N2C)=O
Molecular Weight (g/mol) 728.79
Formula Weight 728.79
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease